Assay Details
| Gene Symbol: DNAJC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710248.3 | XP_002710294.1 | 3 - 4 | 446 | 80 | |
| XM_008255105.2 | XP_008253327.1 | 3 - 4 | 448 | 80 | ||
| XM_017341088.1 | XP_017196577.1 | 3 - 4 | 381 | 80 | ||
| XM_017341089.1 | XP_017196578.1 | 2 - 3 | 328 | 80 | ||
| XM_017341090.1 | XP_017196579.1 | 3 - 4 | 429 | 80 | ||
More Information
Set Membership: |
|

DNAJC18
Genomic Map