Assay Details
| Gene Symbol: TEX35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263812.2 | XP_008262034.2 | 7 - 8 | 716 | 60 | |
| XM_017345636.1 | XP_017201125.1 | 7 - 8 | 662 | 60 | ||
| XM_017345637.1 | XP_017201126.1 | 6 - 7 | 557 | 60 | ||
| XM_017345638.1 | XP_017201127.1 | 7 - 8 | 704 | 60 | ||
More Information
Set Membership: |
|

TEX35
Genomic Map