Assay Details
| Gene Symbol: NINJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259849.2 | XP_008258071.1 | 2 - 3 | 584 | 67 | |
| XM_017343444.1 | XP_017198933.1 | 2 - 3 | 1077 | 67 | ||
| XM_017343445.1 | XP_017198934.1 | 2 - 3 | 542 | 67 | ||
| XM_017343446.1 | XP_017198935.1 | 3 - 4 | 412 | 67 | ||
More Information
Set Membership: |
|

NINJ2
Genomic Map