Assay Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261621.2 | XP_008259843.1 | 1 - 2 | 132 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008261621.2 transcripts.
Set Membership: |
|

KBTBD2
Genomic Map