Assay Details
| Gene Symbol: TEX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252276.2 | XP_008250498.1 | 9 - 10 | 9738 | 65 | |
| XM_008252281.2 | XP_008250503.1 | 10 - 11 | 9737 | 65 | ||
| XM_008252288.2 | XP_008250510.1 | 9 - 10 | 9713 | 65 | ||
| XM_008252299.2 | XP_008250521.1 | 10 - 11 | 9729 | 65 | ||
More Information
Set Membership: |
|

TEX15
Genomic Map