Assay Details
| Gene Symbol: CCDC50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266659.2 | XP_008264881.1 | 2 - 3 | 404 | 62 | |
| XM_008266660.2 | XP_008264882.1 | 2 - 3 | 398 | 62 | ||
| XM_008266662.2 | XP_008264884.1 | 2 - 3 | 407 | 62 | ||
| XM_008266663.2 | XP_008264885.1 | 2 - 3 | 418 | 62 | ||
| XM_008266664.2 | XP_008264886.1 | 2 - 3 | 388 | 62 | ||
| XM_017346992.1 | XP_017202481.1 | 3 - 4 | 467 | 62 | ||
More Information
Set Membership: |
|

CCDC50
Genomic Map