Assay Details
| Gene Symbol: LAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273154.2 | XP_008271376.1 | 7 - 8 | 1090 | 84 | |
| XM_008273155.2 | XP_008271377.1 | 7 - 8 | 1090 | 84 | ||
| XM_008273156.2 | XP_008271378.1 | 7 - 8 | 1090 | 84 | ||
More Information
Set Membership: |
|

LAMP2
Genomic Map