Assay Details
| Gene Symbol: IL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255687.2 | XP_008253909.1 | 5 - 6 | 505 | 82 | |
| XM_008255688.2 | XP_008253910.1 | 4 - 5 | 2879 | 82 | ||
More Information
Set Membership: |
|

IL7
Genomic Map