Assay Details
| Gene Symbol: FBXO43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710712.3 | XP_002710758.1 | 2 - 3 | 2265 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXO43| Gene Symbol: FBXO43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710712.3 | XP_002710758.1 | 2 - 3 | 2265 | 66 | |
Set Membership: |
|