Assay Details
| Gene Symbol: FIGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721581.3 | XP_002721627.1 | 2982 | 80 | ||
| XM_008248428.2 | XP_008246650.1 | 2677 | 80 | |||
More Information
Set Membership: |
|

FIGNL1
Genomic Map