Assay Details
| Gene Symbol: MTERF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257104.2 | XP_008255326.1 | 1 - 2 | 515 | 85 | |
| XM_008257106.2 | XP_008255328.1 | 1 - 2 | 519 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008257104.2, XM_008257106.2 transcripts.
Set Membership: |
|

MTERF2
Genomic Map