Assay Details
| Gene Symbol: MTERF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257104.2 | XP_008255326.1 | 2 - 3 | 627 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008257104.2 transcripts.
Set Membership: |
|

MTERF2
Genomic Map