Assay Details
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721340.3 | XP_002721386.1 | 511 | 69 | ||
| XM_008275046.2 | XP_008273268.1 | 384 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNX20| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721340.3 | XP_002721386.1 | 511 | 69 | ||
| XM_008275046.2 | XP_008273268.1 | 384 | 69 | |||
Set Membership: |
|