Assay Details
| Gene Symbol: BBS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256822.2 | XP_008255044.1 | 2 | 3620 | 134 | |
| XM_008256823.2 | XP_008255045.1 | 2 | 3494 | 134 | ||
| XM_008256824.2 | XP_008255046.1 | 2 | 3479 | 134 | ||
More Information
Set Membership: |
|

BBS10
Genomic Map