Assay Details
| Gene Symbol: ZNF711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720149.3 | XP_002720195.1 | 4 - 5 | 837 | 78 | |
| XM_008272987.2 | XP_008271209.1 | 5 - 6 | 1586 | 78 | ||
| XM_008272988.2 | XP_008271210.1 | 6 - 7 | 1214 | 78 | ||
| XM_008272990.2 | XP_008271212.1 | 5 - 6 | 958 | 78 | ||
| XM_008272991.2 | XP_008271213.1 | 4 - 5 | 1514 | 78 | ||
| XM_017349880.1 | XP_017205369.1 | 2 - 3 | 2189 | 78 | ||
| XM_017349881.1 | XP_017205370.1 | 3 - 4 | 734 | 78 | ||
| XM_017349882.1 | XP_017205371.1 | 3 - 4 | 762 | 78 | ||
More Information
Set Membership: |
|

ZNF711
Genomic Map