Assay Details
| Gene Symbol: SH3RF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255168.2 | XP_008253390.1 | 3 - 4 | 713 | 67 | |
| XM_008255169.2 | XP_008253391.1 | 3 - 4 | 792 | 67 | ||
| XM_017341158.1 | XP_017196647.1 | 2 - 3 | 632 | 67 | ||
| XM_017341159.1 | XP_017196648.1 | 2 - 3 | 965 | 67 | ||
More Information
Set Membership: |
|

SH3RF2
Genomic Map