Assay Details
| Gene Symbol: HSD17B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257668.2 | XP_008255890.1 | 2 - 3 | 756 | 71 | |
| XM_017342390.1 | XP_017197879.1 | 2 - 3 | 766 | 71 | ||
More Information
Set Membership: |
|

HSD17B2
Genomic Map