Assay Details
| Gene Symbol: SEC61G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713870.3 | XP_002713916.1 | 3 - 4 | 340 | 95 | |
| XM_017344350.1 | XP_017199839.1 | 3 - 4 | 232 | 95 | ||
More Information
Set Membership: |
|

SEC61G
Genomic Map