Assay Details
| Gene Symbol: LRRC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346108.1 | XP_017201597.1 | 1 | 85 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017346108.1 transcripts.
Set Membership: |
|

LRRC8B
Genomic Map