Assay Details
| Gene Symbol: CDCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260498.2 | XP_008258720.2 | 2 - 3 | 697 | 64 | |
| XM_017343758.1 | XP_017199247.1 | 2 - 3 | 697 | 64 | ||
| XM_017343759.1 | XP_017199248.1 | 2 - 3 | 697 | 64 | ||
More Information
Set Membership: |
|

CDCP1
Genomic Map