Assay Details
Gene Symbol: FAM107B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008268119.2 | XP_008266341.1 | 2 - 3 | 286 | 76 | |
XM_008268120.2 | XP_008266342.1 | 3 - 4 | 372 | 76 | ||
XM_008268121.2 | XP_008266343.1 | 4 - 5 | 345 | 76 | ||
XM_008268122.2 | XP_008266344.1 | 2 - 3 | 274 | 76 | ||
XM_017347691.1 | XP_017203180.1 | 3 - 4 | 281 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
