Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715245.3 | XP_002715291.1 | 5 - 6 | 867 | 107 | |
| XM_008264168.2 | XP_008262390.1 | 5 - 6 | 722 | 107 | ||
| XM_008264169.2 | XP_008262391.1 | 4 - 5 | 701 | 107 | ||
More Information
Set Membership: |
|

FCRLB
Genomic Map