Assay Details
Gene Symbol: PTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008261357.2 | XP_008259579.1 | 2 - 3 | 242 | 63 | |
GenBank mRNA | KP657517.1 | - | 2 - 3 | 168 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
