Assay Details
| Gene Symbol: SH3PXD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255391.2 | XP_008253613.1 | 11 - 12 | 1294 | 67 | |
| XM_017341258.1 | XP_017196747.1 | 12 - 13 | 1378 | 67 | ||
More Information
Set Membership: |
|

SH3PXD2B
Genomic Map