Assay Details
| Gene Symbol: TNNI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268598.2 | XP_008266820.1 | 4 - 5 | 146 | 47 | |
| XM_008268601.1 | XP_008266823.1 | 5 - 6 | 172 | 47 | ||
| XM_017347867.1 | XP_017203356.1 | 4 - 5 | 101 | 47 | ||
More Information
Set Membership: |
|

TNNI1
Genomic Map