Assay Details
| Gene Symbol: GTPBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716701.3 | XP_002716747.2 | 2 - 3 | 555 | 99 | |
| XM_002716702.3 | XP_002716748.2 | 3 - 4 | 645 | 99 | ||
| XM_008266941.2 | XP_008265163.1 | 3 - 4 | 654 | 99 | ||
More Information
Set Membership: |
|

GTPBP8
Genomic Map