Assay Details
| Gene Symbol: TSPAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720923.3 | XP_002720969.1 | 449 | 67 | ||
| XM_008274227.2 | XP_008272449.1 | 785 | 67 | |||
| XM_008274228.2 | XP_008272450.1 | 350 | 67 | |||
More Information
Set Membership: |
|

TSPAN31
Genomic Map