Assay Details
Gene Symbol: CTDSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712475.3 | XP_002712521.1 | 6 - 7 | 800 | 72 | |
XM_017343112.1 | XP_017198601.1 | 6 - 7 | 988 | 72 | ||
XM_017343113.1 | XP_017198602.1 | 6 - 7 | 961 | 72 | ||
XM_017343114.1 | XP_017198603.1 | 6 - 7 | 850 | 72 | ||
XM_017343115.1 | XP_017198604.1 | 6 - 7 | 768 | 72 | ||
XM_017343116.1 | XP_017198605.1 | 6 - 7 | 847 | 72 | ||
XM_017343117.1 | XP_017198606.1 | 6 - 7 | 834 | 72 | ||
XM_017343118.1 | XP_017198607.1 | 6 - 7 | 805 | 72 | ||
XM_017343119.1 | XP_017198608.1 | 6 - 7 | 768 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
