Assay Details
| Gene Symbol: SHISA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713345.3 | XP_002713391.1 | 5 - 6 | 738 | 55 | |
| XM_008260607.2 | XP_008258829.1 | 3 - 4 | 435 | 55 | ||
| XM_017343801.1 | XP_017199290.1 | 4 - 5 | 654 | 55 | ||
More Information
Set Membership: |
|

SHISA5
Genomic Map