Assay Details
| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721341.3 | XP_002721387.1 | 385 | 88 | ||
| XM_017350883.1 | XP_017206372.1 | 840 | 88 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNEP1R1| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721341.3 | XP_002721387.1 | 385 | 88 | ||
| XM_017350883.1 | XP_017206372.1 | 840 | 88 | |||
Set Membership: |
|