Assay Details
| Gene Symbol: ZCCHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713921.3 | XP_002713967.1 | 2 - 3 | 490 | 55 | |
| XM_008261882.2 | XP_008260104.1 | 2 - 3 | 473 | 55 | ||
| XM_008261883.2 | XP_008260105.1 | 2 - 3 | 894 | 55 | ||
More Information
Set Membership: |
|

ZCCHC9
Genomic Map