Assay Details
| Gene Symbol: ART5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349915.1 | XP_017205404.1 | 1 - 2 | 862 | 67 | |
| XM_017349916.1 | XP_017205405.1 | 1 - 2 | 865 | 67 | ||
| XM_017349919.1 | XP_017205408.1 | 1 - 2 | 868 | 67 | ||
| XM_017349921.1 | XP_017205410.1 | 1 - 2 | 865 | 67 | ||
More Information
Set Membership: |
|

ART5
Genomic Map