Assay Details
| Gene Symbol: ABHD17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708204.3 | XP_002708250.1 | 4 | 2350 | 76 | |
| XM_008256380.2 | XP_008254602.1 | 6 | 2691 | 76 | ||
| XM_008256385.2 | XP_008254607.1 | 5 | 2497 | 76 | ||
| XM_017341828.1 | XP_017197317.1 | 4 | 1847 | 76 | ||
More Information
Set Membership: |
|

ABHD17B
Genomic Map