Assay Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263384.2 | XP_008261606.1 | 3 - 4 | 439 | 63 | |
| XM_008263385.2 | XP_008261607.1 | 3 - 4 | 397 | 63 | ||
More Information
Set Membership: |
|

FAM229B
Genomic Map