Assay Details
| Gene Symbol: MFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723539.3 | XP_002723585.1 | 850 | 64 | ||
| XM_017339613.1 | XP_017195102.1 | 924 | 64 | |||
| XM_017339614.1 | XP_017195103.1 | 388 | 64 | |||
More Information
Set Membership: |
|

MFNG
Genomic Map