Assay Details
| Gene Symbol: MORN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719734.3 | XP_002719780.1 | 3 - 4 | 637 | 59 | |
| XM_008272193.2 | XP_008270415.1 | 2 - 3 | 536 | 59 | ||
| XM_008272195.2 | XP_008270417.1 | 3 - 4 | 1126 | 59 | ||
More Information
Set Membership: |
|

MORN3
Genomic Map