Assay Details
| Gene Symbol: TMEM14C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714171.3 | XP_002714217.1 | 4 - 5 | 373 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM14C| Gene Symbol: TMEM14C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714171.3 | XP_002714217.1 | 4 - 5 | 373 | 63 | |
Set Membership: |
|