Assay Details
| Gene Symbol: ETV7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262837.2 | XP_008261059.1 | 4 - 5 | 687 | 103 | |
| XM_017345070.1 | XP_017200559.1 | 4 - 5 | 685 | 103 | ||
More Information
Set Membership: |
|

ETV7
Genomic Map