Assay Details
| Gene Symbol: ETV7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262837.2 | XP_008261059.1 | 5 - 6 | 825 | 61 | |
| XM_017345070.1 | XP_017200559.1 | 5 - 6 | 823 | 61 | ||
| XM_017345071.1 | XP_017200560.1 | 3 - 4 | 543 | 61 | ||
More Information
Set Membership: |
|

ETV7
Genomic Map