Assay Details
| Gene Symbol: KBTBD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713083.3 | XP_002713129.2 | 4 - 5 | 1757 | 72 | |
| XM_008260321.1 | XP_008258543.1 | 4 - 5 | 1690 | 72 | ||
| XM_008260322.1 | XP_008258544.1 | 5 - 6 | 1826 | 72 | ||
| XM_008260323.2 | XP_008258545.1 | 5 - 6 | 1892 | 72 | ||
More Information
Set Membership: |
|

KBTBD12
Genomic Map