Assay Details
| Gene Symbol: ZFP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711496.3 | XP_002711542.2 | 5 | 2259 | 102 | |
| XM_008257223.2 | XP_008255445.1 | 5 | 2257 | 102 | ||
| XM_008257225.2 | XP_008255447.1 | 6 | 2335 | 102 | ||
More Information
Set Membership: |
|

ZFP30
Genomic Map