Assay Details
| Gene Symbol: LAYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708467.3 | XP_002708513.1 | 3 - 4 | 558 | 57 | |
| XM_008261598.2 | XP_008259820.1 | 2 - 3 | 532 | 57 | ||
| XM_008261607.2 | XP_008259829.1 | 3 - 4 | 558 | 57 | ||
More Information
Set Membership: |
|

LAYN
Genomic Map