Assay Details
| Gene Symbol: ZNF329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721895.3 | XP_002721941.1 | 2675 | 98 | ||
| XM_008249009.2 | XP_008247231.1 | 2656 | 98 | |||
More Information
Set Membership: |
|

ZNF329
Genomic Map