Assay Details
| Gene Symbol: RAB5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719392.3 | XP_002719438.1 | 4 - 5 | 648 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB5C| Gene Symbol: RAB5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719392.3 | XP_002719438.1 | 4 - 5 | 648 | 69 | |
Set Membership: |
|