Assay Details
| Gene Symbol: YIPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714693.3 | XP_002714739.1 | 8 - 9 | 1148 | 78 | |
| XM_017345150.1 | XP_017200639.1 | 7 - 8 | 1016 | 78 | ||
More Information
Set Membership: |
|

YIPF3
Genomic Map