Assay Details
| Gene Symbol: OSM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274704.2 | XP_008272926.1 | 1478 | 89 | ||
| XM_017350693.1 | XP_017206182.1 | 1972 | 89 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: OSM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274704.2 | XP_008272926.1 | 1478 | 89 | ||
| XM_017350693.1 | XP_017206182.1 | 1972 | 89 | |||
Set Membership: |
|