Assay Details
| Gene Symbol: TNFRSF13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275159.2 | XP_008273381.1 | 164 | 107 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF13C| Gene Symbol: TNFRSF13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275159.2 | XP_008273381.1 | 164 | 107 | ||
Set Membership: |
|