Assay Details
| Gene Symbol: SPCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709480.3 | XP_002709526.1 | 5 | 3585 | 84 | |
| XM_017338296.1 | XP_017193785.1 | 5 | 3540 | 84 | ||
More Information
Set Membership: |
|

SPCS3
Genomic Map