Assay Details
| Gene Symbol: SPRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017340722.1 | XP_017196211.1 | 5 - 6 | 705 | 71 | |
| XM_017340723.1 | XP_017196212.1 | 5 - 6 | 699 | 71 | ||
More Information
Set Membership: |
|

SPRED2
Genomic Map