Assay Details
| Gene Symbol: FAM24A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250246.2 | XP_008248468.1 | 460 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008250246.2 transcripts.
Set Membership: |
|

FAM24A
Genomic Map